missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Support Pack Plus
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
Pack of 10
Description
Includes
PBS (20x)
Assay Buffer (20x)
Wash Buffer (20x)
Substrate Solution
Stop Solution
plates
Specifications
Specifications
| Chemical Name or Material | Assay Buffer, Solution, Wash Buffer |
| For Use With (Application) | ELISA |
| Physical Form | Liquid |
| Quantity | 10 x 96 Tests |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction