missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Support Pack Standard
10 x 96 tests
Brand: Invitrogen™ BMS412
152.67 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Includes:
PBS (20x)
Assay Buffer (20x)
Wash Buffer (20x)
Substrate Solution
Stop Solution
Description
Specifications
10 x 96 Tests | |
ELISA | |
Mouse, Rat, Human | |
Assay Buffer, Wash Buffer |
For Research Use Only. Not for use in diagnostic procedures.