missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Support Stands for Thermo Scientific™ Heratherm Ovens and Incubators

Support stands with casters

£399.22 - £494.77

Specifications

Type Support Stand
Includes Caster
View More Specs

Products 3
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
10811311
View Documents Promotion Details
Thermo Scientific™
50127741
60 L Model Ovens and Incubators
£399.22
Each
In Stock
to see stock.
Add to basket
 
10304130
View Documents Promotion Details
Thermo Scientific™
50127742
100 L Model Ovens and Incubators
£425.49
Each
In Stock
to see stock.
Add to basket
 
10724602
View Documents Promotion Details
Thermo Scientific™
50127743
180 L Model Ovens and Incubators
£494.77
Each
Estimated Shipment: 17-06-2025
to see stock.
Add to basket
 
Description

Description

Support stands with casters

Height including casters: 7.36 in. (18.7cm).

Specifications

Specifications

Support Stand
Caster
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Support Stands for Thermo Scientific™ Heratherm Ovens and Incubators

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.