missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microflex™ High Risk Disposable Nitrile Gloves
13.32 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Extended cuff provides additional protection for the wrist and arm for hazardous environments.
- Advanced nitrile formulation provides the lowest acceptable pinhole rates (AQL 0.65)
- Non-stick properties are built into the glove formulation to create a resistance to tape or adhesives, and to assist workers with improved efficiency and speed.
- Textured fingers ensure strong, efficient grip and excellent worker protection.
- Constructed without natural rubber latex materials or powder to help protect from allergies, skin irritation and dryness.
- Approved for use with chemotherapy drugs
Specifications
Certifications/Compliance | CE, ISO 11193-1, EN420, 89/686/EEC, EN374-2: 2003, EN374-3: 2003, MDD 93/42/EEC, Class 1, EN455-1, EN455-2 and EN455-3, ASTM F1671 |
Color | Blue |
Disposable | Single-use |
Grade | Medical |
Hand | Ambidextrous |
Latex Free | Yes |
Material | Nitrile |
Powdered | Powder Free |
Size | Large |
Sterility | Non-sterile |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'