Learn More
Invitrogen™ SureLock™ Tandem Midi Gel Tank
The Invitrogen SureLock Tandem Midi Gel Tank provides for rapid electrophoresis of midi gels using minimal buffer (∼520 mL/gel) in a leak-free system. With a setup time of ∼30 seconds, the tank efficiently runs midi gels while also providing consistent performance.
Brand: Invitrogen™ STM1001
Description
The tank features two independent chambers, allowing electrophoresis of one or two gels at a time, which saves on buffer and limits waste. The tank is compatible with all Invitrogen pre-cast midi gels and midi gel cassettes.
Features:
- 2-in-1 gel electrophoresis and transfer tank—enables electrophoresis and transfer of high-performance Invitrogen midi gels using the same tank
- Two separate chambers—run 1 or 2 gels or transfers and use only the necessary amount of buffer for each gel, minimizing buffer cost and waste
- Optimal performance with fast protocols—saves time while yielding better results
- Room temperature transfer when paired with SureLock Tandem Midi Blot Modules—eliminates the need to pre-chill buffers and the hassle and messiness of ice baths
- User-friendly—easy setup and simple workflow
- Does not require dummy cassette or buffer dam—reduces accessories
- Compatible—fits all Invitrogen precast midi gels and midi gel cassettes
- Modern design provides easy access to sample loading area reducing sample loading time and effort
- Robust design using strong polycarbonate plastic that prevents cracks and leaks
- Unobstructed visibility to your gels as they run
- Easy fitting lid with guiding notches and wire management hooks
The SureLock Tandem Midi Gel Tank can be used for wet tank transfers when paired with the SureLock Tandem Midi Blot Module. The SureLock Tandem Midi Blot Module performs efficient, room-temperature, wet protein transfers for downstream western blot analysis. The tank accommodates two blot modules, allowing transfer of one or two gels at a time. The unique design of the SureLock Tandem Blot Module uses considerably less transfer buffer (only ∼300 mL per transfer) than other wet transfer systems. This lower buffer requirement keeps the amount of methanol waste (a hazardous material) to a minimum.
Specifications
Contains: • 1 SureLock Tandem Midi Gel Tank • 1 Gel Knife |
|
Midi | |
Midi Gel Tank | |
Gel size: 8 x 13 cm (Midi or Wide Format Gels)Gel cassette: 10.3 x 15 cmThickness: 1.0 mm | |
Room Temperature | |
25 x 17.9 x 17.3 cm (height with lid on) |
All Invitrogen precast midi gels, and Invitrogen empty midi gel cassettes | |
SureLock | |
All Invitrogen Precast Midi Gels, and Invitrogen Empty Midi Gel Cassettes | |
1 | |
One year |