missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laboratoires Anios™ Surfanios Premium
Cleaning and disinfection of floors, walls, medical equipment and non-invasive medical devices.
Specifications
Product Type | Surfanios Premium |
---|---|
Composition | N- (3- aminopropyl) -N- dodecylpropane -1,3- diamine, didecyldimethylammonium chloride, excipients |
For Use With (Application) | For Cleaning and Disinfection of Floors and Surfaces |
Description
Detergent disinfectant for floors and surfaces:
- Green coloured solution
- pH of pure product: 11.2
- pH of diluted product (0.25%): approx 8
- Density: 1.007
- Refraction index: 1.3587
Specifications
Surfanios Premium | |
N- (3- aminopropyl) -N- dodecylpropane -1,3- diamine, didecyldimethylammonium chloride, excipients | |
For Cleaning and Disinfection of Floors and Surfaces |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- SURFANIOS PREMIUM
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P102-Keep out of reach of children.
- P234-Keep only in original container.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : N-(3-AMINOPROPYL)-N-DODECYLPROPANE-1, 3-DIAMINE, DIDECYLDIMETHYLAMMONIUM CHLORIDE, PROPAN-2-OL
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title