missing translation for 'onlineSavingsMsg'
Learn More
Kolmi Hopen™ Surgical Patient Gown with Back Laces Product Code.: 17720169
Add to basket

Kolmi Hopen™ Surgical Patient Gown with Back Laces

Product Code. 17720169
Polypropylene Patient Gown with Back Laces; Closure Type: Tie; Color: Navy Blue; For Use With (Application): Medical; Disposable: Yes; Size: Universal
Click to view available options
Polypropylene Patient Gown with Back Laces; Closure Type: Tie; Color: Navy Blue; For Use With (Application): Medical; Disposable: Yes; Size: Universal
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Product Code. 17720169

Brand: Kolmi Hopen™ J40460

Add to basket
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Lightweight patient gown with long sleeves and tie closure in back.

  • Light and breathable surgical shirt
  • Collarless model
  • Dorsal closure with laces
  • Higher weight than other medical gowns, guaranteeing better opacity

Specifications

Size Universal
For Use With (Application) Medical
Gender Unisex
No. of Pockets None
Product Type General Purpose Gown
Material Polypropylene
Color Navy
Closure Type Ties
Disposable Single-use
Sterility Non-sterile
Weight (Metric) 42 g/m2
Autoclavable Not Autoclavable
No. per Pack 10 Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kolmi Hopen™ Surgical Patient Gown with Back Laces >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.