Learn More
Poulten Selfe™ Suspended-Level Shortened Form Glass Viscometer
Brand:  Poulten Selfe™ 1628/02
Product Code. 13161053
  • £198.00 / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
2.1v to 3v(mm2s to 1,cst) with certificate of calibration Poulten Selfe and Lee