Learn More
SVL™ Bearing Stirrer for SVL™ Stirrer Shaft
Designed to accept SVL™ stirrer shafts with ground glass surfaces.
Brand:  SVL 588-09
 View more versions of this product
Product Code. 11937298
  • £239.00 / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Borosilicate glass complete with fittings
  • Glands manufactured from PTFE based polymer which is self lubricating and corrosion-resistant
  • Two types available: With SVL™ screwthread to allow connection to SVL™ flasks, With Quickfit™ ground glass cone to allow connection to Quickfit™ flasks
TRUSTED_SUSTAINABILITY
Specifications
Specifications
29/32
Borosilicate Glass