Learn More
SVL™ Cone/Screw Thread Adapter
Product Code. p-8003028
Click to view available options
Cone Size:
14/23
24/29
Unit Size:
Pack of 10
Product Code. 15577740
Brand: SVL 67070
Only null left
Add to basket
Add to basket
Facilitates connection between SVL™ and Quickfit™ glassware.
  • Features an SVL™ screwthread and Quickfit™ ground glass socket
  • Thread accepts SVL™ screw caps and couplings. Cone fits Quickfit™ sockets
  • Manufactured from temperature and chemical resistant borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
Cone Size 14/23
For Use With (Equipment) SVL™ and Quickfit™ Glassware
Material Borosilicate Glass
Product Type Cone/Screw thread adapter
Standard Taper Joint No. 15