missing translation for 'onlineSavingsMsg'
Learn More

Rotulex™ Metal Joint Clamps

Spring-loaded metal clips. Rotulex™ Metal Joint Clamps are designed to secure corresponding Rotulex™ glassware systems with ball and socket style joints.

Brand:  SVL RC29

27.48 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12099111

  • £39.05 / Each
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Suitable for high temperature work
  • System includes male ball joint, PTFE covered O-ring and female cup joint which are secured with the joint clamp
  • Components are ordered separately
  • Available in a variety of sizes
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Joint Clamp
Metal
Rotulex™ spherical joint system
Approx. bore size: 15 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers