missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rotulex™ Metal Joint Clamps
Spring-loaded metal clips. Rotulex™ Metal Joint Clamps are designed to secure corresponding Rotulex™ glassware systems with ball and socket style joints.
Brand: SVL RC35
44.79 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Suitable for high temperature work
- System includes male ball joint, PTFE covered O-ring and female cup joint which are secured with the joint clamp
- Components are ordered separately
- Available in a variety of sizes
Specifications
Joint Clamp | |
Metal | |
Rotulex™ spherical joint system | |
Approx. bore size: 20 mm |