Learn More
SVL™ Sealing Rings for Sliding Joints
Manufactured from silicone rubber and sheathed with PTFE for protection against chemical erosion. SVL™ Sealing Rings for Sliding Joints are ideal for inserting thermometers and glass tubing into an SVL™ system. A wide range of sizes is available for use with tubing from 4.0 to 32.8mm.
£63.95 - £296.00
Specifications
Material | Silicone Rubber, PTFE (Sheath) |
---|---|
Type | Sealing Ring |
For Use With (Application) | Torion joints and sliding joints |
Product Code | Brand | Description | Joint Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Joint Size | Price | Quantity & Availability | ||||
10105674
|
SVL
701-22 |
Fits Tubing O.D.: 6.6 to 7.4mm | 15 |
£83.30
Pack of 10 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
12517816
|
SVL
701-56 |
Fits Tubing O.D.: 17.6 to 18.4mm | 30 |
£160.00
Pack of 10 |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
10287774
|
SVL
701-29 |
Fits Tubing O.D.: 31.2 to 32.8mm | 42 |
£296.00
Pack of 10 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
12607245
|
SVL
701-24 |
Fits Tubing O.D.: 13.6 to 14.4mm | 22 |
£150.00
Pack of 10 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||||
10739365
|
SVL
701-27 |
Fits Tubing O.D.: 21.5 to 22.5mm | 30 |
£119.00
Pack of 10 |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
10442205
|
SVL
701-23 |
Fits Tubing O.D.: 7.6 to 8.4mm | 15 |
£63.95
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
10635905
|
SVL
701-19 |
Fits Tubing O.D.: 4.0 to 4.4mm | 15 |
£72.55
Pack of 10 |
Estimated Shipment: 23-10-2024
Log in to see stock. |
||||
10645905
|
SVL
701-21 |
Fits Tubing O.D:. 5.6 to 6.4mm | 15 |
£75.30
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
10146444
|
SVL
701-25 |
Fits Tubing O.D.: 15.6 to 16.4mm | 22 |
£107.00
Pack of 10 |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
10319944
|
SVL
701-51 |
Fits Tubing O.D.: 9.6 to 10.4mm | 22 |
£111.00
Pack of 10 |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
10379984
|
SVL
701-52 |
Fits Tubing O.D.: 11.6 to 12.4mm | 22 |
£115.00
Pack of 10 |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
10014464
|
SVL
701-26 |
Fits Tubing O.D.: 19.5 to 20.5mm | 30 |
£122.00
Pack of 10 |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
11943889
|
SVL
701-28 |
Fits Tubing O.D: 29.2 to 30.8mm | 42 |
£296.00
Pack of 10 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||||
Description
- Sealing rings designed to provide a sliding joint
- Ideal for inserting thermometers and glass tubing etc. into an SVL™ system
- Sealing rings manufactured from silicone rubber and sheathed with PTFE for protection against chemical erosion
- Wide range of sizes for use with tubing from 4.0 to 32.8mm
Specifications
Silicone Rubber, PTFE (Sheath) | |
Sealing Ring | |
Torion joints and sliding joints |