Learn More
Eppendorf™ Swing Bucket Rotor for Benchtop Centrifuge
Product Code. 10394792 Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
A-4-38
A-4-44
A-4-62
A-4-62-MTP
A-4-81
A-4-81-MTP
A-8-17
Includes:
4 Hangers for 2 x 50 mL Falcon-G
4 MTP buckets
4 x 100mL rectangular buckets
4 x 250mL rectangular buckets
4 x 500mL rectangular buckets
4 x 85mL buckets
8 x 15mL buckets
Unit Size:
Each
Product Code. 10394792
Brand: Eppendorf™ 5810711002
Use these rotors for a wide range of molecular applications. Eppendorf™ Swing Bucket Rotors are designed for use with Eppendorf benchtop centrifuges.
  • Accommodates tubes and plates
  • Aerosol-tight caps for safe centrifugation of hazardous samples optionally available
  • Rotor life time: max. 40,000 cycles
  • Rotor, buckets, caps and adapters are autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Type Swinging Bucket Rotor
For Use With (Equipment) A-4-62-MTP
Includes 4 MTP buckets