Learn More
BINDER™ Swivel Casters
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB 260, KB 470, KB 720, KB 1060
KB PRO Series
KBF PRO Series
KBF Series
Unit Size:
Each
Description
Height-adjustable swivel casters for horizontal alignment of unit on uneven floors.
Specifications
Specifications
| For Use With (Equipment) | KB PRO Series |
| Material | Steel |
| Type | Castor Frame |