missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYBR GreenER™ qPCR SuperMix for ABI PRISM™ Instrument

Incorporate the latest high-performance technology to provide the most reliable gene expression data from your ABI 7300, 7000, 7700 or 7900HT instrument.

Brand:  Invitrogen™ 11760100

131.00 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10512773

  • £262.00 / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

The SYBR™ GreenER™ qPCR SuperMix kits include a dsDNA-binding dye that produces a brighter signal and significantly less PCR inhibition, for improved qPCR performance over a broad dynamic range.

Consistent Specificity and Low-Copy Detection: The SYBR™ GreenER™ qPCR reagent system minimizes the formation of nonspecific products, including primer-dimers, for greater accuracy across a wide range of targets. The reliability of the SYBR™ GreenER™ qPCR reagent system also translates to greater sensitivity. With SYBR™ GreenER™ qPCR SuperMix, you can consistently detect fewer than 10 copies of a target in genomic DNA.

  • The SYBR™ GreenER™ system is specially formulated to offer the best sensitivity, specificity, and reproducibility
  • A novel dsDNA-binding dye - exhibits a brighter signal for increased sensitivity and less PCR inhibition than original SYBR™ Green I, while using the same instrument filters and settings
  • An optimized buffer system - improves sensitivity and provides excellent long-term stability
  • Uracil DNA glycosylase - reduces carryover contamination in qPCR

Order Info

Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

SYBR™ GreenER™ qPCR SuperMix for ABI PRISM™ instruments contains:

2X SYBR™ GreenER™ qPCR SuperMix premixed with ROX reference dye.

Store at +4°C upon receipt. Guaranteed stable for 6 months when properly stored.
High
Fast or Standard
Platinum™, SYBR™ GreenER™
Dry Ice
Gene Expression
100 Reactions
SYBR
qPCR
7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System
Real Time PCR SYBR Master Mix
100 Reactions
2X
DNA (Genomic), cDNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.