missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Syk Inhibitor

Product Code. 15446299
Click to view available options
Quantity:
1 mg
5 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

A cell-permeable oxindole compound that acts as a potent, reversible, and ATP-competitive Syk inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622387-85-3
Molecular Formula C18H15N3O3S
Synonym Spleen Tyrosine Kinase Inhibitor

Specifications

Quantity 5 mg
Description Dark orange
Content And Storage 2°C to 8°C
For Use With (Application) A cell-permeable oxindole compound that acts as a potent, reversible, and ATP-competitive Syk inhibitor
Formula Weight 353.40
Sensitivity Light Sensitive
Solubility Soluble in DMSO
Form Solid
InChI Key MLKHXLFEYOOYEY-UHFFFAOYSA-N
SMILES CN1C=C(C=C2C(=O)NC3=C2C=C(C=C3)S(N)(=O)=O)C2=CC=CC=C12
IUPAC Name 3-[(1-methyl-1H-indol-3-yl)methylidene]-2-oxo-2,3-dihydro-1H-indole-5-sulfonamide
Molecular Weight (g/mol) 353.40
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'