Learn More
sym-Diphenylcarbazide, 98%, ACS reagent, Thermo Scientific Chemicals
£32.90 - £331.00
Chemical Identifiers
CAS | 140-22-7 |
---|---|
Molecular Formula | C13H14N4O |
Molecular Weight (g/mol) | 242.28 |
InChI Key | KSPIHGBHKVISFI-UHFFFAOYSA-N |
Synonym | 1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl |
PubChem CID | 8789 |
ChEBI | CHEBI:4641 |
IUPAC Name | 1,3-dianilinourea |
SMILES | C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10427000
|
Thermo Scientific Acros
158840250 |
25 g | Glass bottle |
£32.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10437000
|
Thermo Scientific Acros
158841000 |
100 g | Glass bottle |
£88.20
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11925661
|
Thermo Scientific Acros
158845000 |
500 g | Glass bottle |
£331.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
140-22-7 | |
242.28 | |
1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl | |
CHEBI:4641 | |
C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2 |
C13H14N4O | |
KSPIHGBHKVISFI-UHFFFAOYSA-N | |
8789 | |
1,3-dianilinourea |
Specifications
140-22-7 | |
Beige to Pink | |
Authentic | |
C13H14N4O | |
(to chromate) passes test | |
1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl | |
KSPIHGBHKVISFI-UHFFFAOYSA-N | |
1,3-dianilinourea | |
8789 | |
242.28 | |
ACS Reagent | |
sym-Diphenylcarbazide |
173.0°C to 176.0°C | |
1% max. | |
97.5 % min. (HPLC) | |
15,292 | |
15,3352 | |
Solubility in water: slightly soluble. Other solubilities: soluble in glacial acetic acid,soluble in alcohol and acetone | |
C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2 | |
242.28 | |
CHEBI:4641 | |
98% | |
Powder or Flakes |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective g
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 205-403-7
- sym-Diphenylcarbazide
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only