Learn More
SymLine Tubes, FlexTube & FlexTube Pro
The two most flexible and adaptable tubing systems for the safe disposal of liquid HPLC waste - flexible, expandable, chemically resistant PTFE tubing with a large bending radius.
Brand: SymLine 106575
113.45 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Additional Details : Weight : 0.70000kg
Description
Specifications
FlexTube Pro | |
PTFE-EC | |
23.622 in. | |
Black |
Symline FlexTube Pro, GL25 (f), ID = 13,4 - 13,9 mm, OD = 16,0 - 16,9 mm outside spiral formed and inside smooth, PTFE electrostatic conductive, Length 600 mm - flexible adaptable | |
16.9 mm | |
600 mm |