missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation Symmetry C18 Column

Symmetry C18 columns feature monofunctionally bonded C18 ligands on a high purity base-deactivated silica.

Brand:  Waters Corporation 186003302

Encompass

 View more versions of this product

Product Code. 13135009

  • £2835.00 / Each
Estimated Shipment: 05-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Symmetry columns have the tightest column specifications in the industry giving resulting highly reproducible chromatographic results.
  • Symmetry columns continues to set the standard for HPLC drug assays, giving you confidence in the long-term compliance of your HPLC methods.
  • No need to compromise on peak shape, selectivity, column-to-column and batch-to-batch reproducibility, or other critical performance characteristics.
  • Available in a variety of particle sizes, chemistries and dimensions.
Specifications

Specifications

40 mm
1500 psi (100 Bar)
7 μm
1/Ea.
Yes
100 mm
Spherical
100 Å
L1
Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers