missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptophysin Monoclonal Antibody (7H12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515623
This item is not returnable.
View return policy
Description
MA5-15623 targets SYP in indirect ELISA, IHC and WB applications and shows reactivity with Human and Rat samples. The MA5-15623 immunogen is purified recombinant fragment of SYP expressed in E. Coli. MA5-15623 detects SYP which has a predicted molecular weight of approximately 33.8kDa.
Synaptophysin is a calcium-binding and integral membrane glycoprotein present in presynaptic vesicles in almost all neurons. Synaptophysin has four transmembrane domains and it forms a complex with dynamin at high calcium concentrations suggesting an involvement in synaptic vesicle endocytosis. It is also involved in the regulation of short-term and long-term synaptic plasticity. Synaptophysin is currently the most widely used marker for nerve terminals and for differentiating neuroendocrine tumors. Mutations in the gene can result in mental retardation, X-linked 96.Specifications
Synaptophysin | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P07825, P08247, Q62277 | |
Syp | |
Purified recombinant fragment of SYP expressed in E. Coli. | |
RUO | |
20977, 24804, 6855 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
7H12 | |
Unconjugated | |
Syp | |
A230093K24Rik; AI848995; BM89 antigen; I79_012252; Major synaptic vesicle protein p38; MRX96; MRXSYP; p38; Syn; Synaptophysin; Syp; Syp I; Syp1 | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction