Learn More
Agilent BioTek Synergy HTX Multi-Mode Microplate Reader, IVD
The BioTek Synergy HTX is a compact, economical multi-mode reader for 6- to 384-well microplates and Take3 microvolume plates. For In Vitro Diagnostic Use.
Brand: Agilent BioTek S1LFTA-SI
Description
The BioTek Synergy HTX is a compact, economical multi-mode reader for 6- to 384-well microplates and Take3 microvolume plates. The unique dual optics design provides superior performance for UV-Vis absorbance, fluorescence, luminescence, and AlphaScreen/AlphaLISA workflows
With incubation and shaking, plus a dual reagent injector module, the system will meet all your assay requirements now and in the future. Synergy HTX is controlled by the easy-to-use, yet powerful, Gen5 software for data collection, analysis, exporting, and reporting.
Specifications
200 to 999 nm, 1 nm increments | |
CE and TUV marked. RoHS compliant. | |
BioStack and Third Party Automation Compatible | |
4-Zone Incubation to 50°C with Condensation Control (±0.2°C at 37°C) | |
Linear, Orbital | |
10 in. | |
25.4 cm | |
S1LFTA | |
Multi-Mode Microplate Reader | |
100/240 V | |
18 kg | |
40.6 cm | |
Photodiode, PMT | |
Xenon Flash, Tungsten Halogen (Alpha Detection/Fluorescence Intensity) |
Absorbance: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD; Dispense: ±1 μL or 2% | |
38 cm | |
15 in. | |
31 sec. (96 wells), 80 sec. (384 wells) | |
Absorbance: 0 to 4 OD; Fluorescence Intensity/Luminesence: >6 decades | |
50/60 Hz | |
Absorbance; Luminescence; Fluorescence (Top/Bottom); Time-resolved Fluorescence (TRF - Secondary Mode); Alpha; Incubation; Shaking | |
2 Syringe Pumps | |
2 | |
40 lb. | |
16 in. | |
6- to 384-well Plates | |
Gen5 Microplate Reader and Imager Software, Gen5 Secure for 21 CFR Part 11 compliance (option) | |
End Point, Kinetic, Spectral Scanning, Well Area Scanning |