Learn More
Agilent BioTek Synergy Neo2 Hybrid Multi-Mode Microplate Reader

BioTek Synergy Neo2 multi-mode microplate reader features patented Hybrid Technology, with independent optical paths that ensure excellent performance in all detection modes. For Research Use Only. Not for use in diagnostic procedures.
POA
Specifications
Absorbance Range | 230 to 999 nm, 1 nm increments |
---|---|
Accuracy | Monochromator: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
CO2 Concentration Range | 0 to 20% |
Depth (Metric) | 52.5 cm |
Product Code | Brand | Model | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Includes | Price | Quantity & Availability | ||||
17288009
![]() |
Agilent BioTek
N2-SN |
NEO2 | Monochromator: ABS; Filter: Dual filter top FI/FP/TRF and LUM |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17248019
![]() |
Agilent BioTek
N2M-SN |
NEO2M | Monochromator: ABS, FI and LUM; Filter: Dual filter top FI/FP/TRF and LUM |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17248029
![]() |
Agilent BioTek
N2MB-SN |
NEO2MB | Monochromater: ABS, FI and LUM; Filter: Dual filter top FI/FP/TRF and LUM, Filter bottom FI/FP/TRF |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17228039
![]() |
Agilent BioTek
N2S-SN |
NEO2S | Monochromater: ABS; Filter: Single filter top FI/FP/TRF and LUM |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17268039
![]() |
Agilent BioTek
N2SM-SN |
NEO2SM | Monochromater: ABS, FI and LUM; Filter: Single filter top FI/FP/TRF and LUM |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17228049
![]() |
Agilent BioTek
N2SMB-SN |
NEO2SMB | Monochromater: ABS, FI and LUM; Filter: Single filter top FI/FP/TRF and LUM, Filter bottom FI/FP/TRF |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17268029
![]() |
Agilent BioTek
N2MBT-SN |
NEO2MBT | Monochromater: ABS, FI and LUM; Filter: Dual filter top FI/FP/TRF and LUM, Filter bottom FI/FP/TRF, TRF Laser |
POA
1 unit |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17288029
![]() |
Agilent BioTek
N2MON-SN |
NEO2MON | Monochromater: ABS, FI and LUM |
POA
1 unit |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17208039
![]() |
Agilent BioTek
N2MT-SN |
NEO2MT | Monochromator: ABS, FI and LUM; Filter: Dual filter top FI/FP/TRF and LUM, TRF Laser |
POA
1 unit |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17248049
![]() |
Agilent BioTek
N2T-SN |
NEO2T | Monochromater: ABS; Filter: Dual filter top FI/FP/TRF and LUM, TRF Laser |
POA
1 unit |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17208019
![]() |
Agilent BioTek
N2ABT-SN |
NEO2ALPHABT | Monochromater: ABS; Filter: Dual filter top FI/FP/TRF and LUM, Filter bottom FI/FP/TRF, Alpha Laser, TRF Laser |
POA
1 unit |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17228019
![]() |
Agilent BioTek
N2AT-SN |
NEO2ALPHAT | Monochromater: ABS; Filter: Dual filter top FI/FP/TRF and LUM, Alpha Laser, TRF Laser |
POA
1 unit |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17228029
![]() |
Agilent BioTek
N2MAT-SN |
NEO2MALPHAT | Monochromater: ABS, FI and LUM; Filter: Dual filter top FI/FP/TRF and LUM, Alpha Laser, TRF Laser |
POA
1 unit |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17208029
![]() |
Agilent BioTek
N2MABT-SN |
NEO2MALPHABT | Monochromater: ABS, FI and LUM; Filter: Dual filter top FI/FP/TRF and LUM, Filter bottom FI/FP/TRF, Alpha Laser, TRF Laser |
POA
1 unit |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17208049
![]() |
Agilent BioTek
N2SMAB-SN |
NEO2SMALPHAB | Monochromater: ABS, FI and LUM; Filter: Single filter top FI/FP/TRF and LUM, Filter bottom FI/FP/TRF, Alpha Laser |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17268019
![]() |
Agilent BioTek
N2MA-SN |
NEO2MALPHA | Monochromater: ABS, FI and LUM; Filter: Dual filter top FI/FP/TRF and LUM, Alpha Laser |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17288019
![]() |
Agilent BioTek
N2MAB-SN |
NEO2MALPHAB | Monochromater: ABS, FI and LUM; Filter: Dual filter top FI/FP/TRF and LUM, Filter bottom FI/FP/TRF, Alpha Laser |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17248039
![]() |
Agilent BioTek
N2SA-SN |
NEO2SALPHA | Monochromater: ABS; Filter: Single filter top FI/FP/TRF and LUM, Alpha Laser |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
17288039
![]() |
Agilent BioTek
N2SMA-SN |
NEO2SMALPHA | Monochromater: ABS, FI and LUM; Filter: Single filter top FI/FP/TRF and LUM, Alpha Laser |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
BioTek Synergy Neo2 multi-mode microplate reader features patented Hybrid Technology, with independent optical paths that ensure excellent performance in all detection modes. Variable bandwidth quad monochromators, sensitive high transmission filter-based optics, laser TRF, and up to 4 PMTs provide ultrafast measurements.
Advanced environmental controls, including CO2/O2 control, incubation to 70°C, and variable shaking support live cell assays. BioStack Neo delivers walk-away automation, high throughput, barcode-labeled filter cubes to streamline workflows and limit errors. Powerful Gen5 software provides complete reader control, powerful data analysis, and LIMS integration.
Specifications
230 to 999 nm, 1 nm increments | |
CE and TUV marked. RoHS compliant. | |
52.5 cm | |
±0.2°C at 37°C | |
0 to 4 OD | |
HTS, biomarker, nucleic acid, protein quantification, rapid kinetics, HTRF, SNP, TR FRET, BRET, Cytokines, ELISA, FP | |
41 cm | |
250 W max. | |
78 lb. | |
15.4 in. | |
Photodiode, Fluorescence/Alpha: Dual PMT or Single PMT (option) | |
Xenon Flash |
Monochromator: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD | |
0 to 20% | |
20.7 in. | |
Linear, Orbital, Double Orbital | |
5 to 1000 μL, 1 μL increments | |
16.1 in. | |
Multi-Mode Microplate Reader | |
0.0001 OD | |
35 kg | |
39 cm | |
Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.