missing translation for 'onlineSavingsMsg'
Learn More

Synoptics™ Gel Imaging DigiGenius
GREENER_CHOICE

DigiGenius is a digital camera with hood UV filter. One copy of GeneTools. Transilluminator converter not supplied. Standard Warranty 1 year. PC required (not included).

Brand:  Synoptics™ DG1

2466.65 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.


Product Code. 12824038

  • £2780.00 / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The DigiGenius offers reproducible, high resolution digital images for gel documentation. The system is easy to use and allows you to capture and view images directly on your PC or MAC. DigiGenius gel documentation system is ideal if you have a limited budget because you can purchase the basic system and then add a UV transilluminator, white light table or conversion screen, safety hood, mini darkroom and printer, as well as Syngene's industry leading GeneTools analysis software, as the need arises.
  • Electrophoresis gels - Ethidium Bromide, Coomassie Blue, Silver Stain, SYBR™ Gold, SYBR™ Green, SYBR™ Safe, GelStar™, SYPRO™ Red, SYPRO™ Ruby, SYPRO™ Orange, Fluorescein, Rhodamine Red™, Texas Red™, Pro-Q™ Diamond , Deep Purple™
  • Agar plates
  • Cells in flasks
  • Autoradiographs
  • Membranes
  • Spots and slot blots
  • Microtitre plates
  • Macro-arrays
  • Low cost gel documentation system
  • Digital camera - high quality images with precise band separation
  • Budget system with analysis software - cost effective method of upgrading as fluorescent applications change
  • Interface to PC or Mac - full control of camera set-up and image capture

Note: A transilluminator needs to be purchased separately.

Specifications

Specifications

None
115 V
210 mm
8.2 in.
14.6 in.
370 mm
Gel Imaging
Gel Documentation System
50 Hz
11 in.
UV
18 MP
370 mm
24 Bit Color
Syngene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers