Learn More
Syngene™ UV Transilluminator 2020M
UV Transilluminator 2020M is for illumination of any sample excited by UV light.
Brand: Syngene™ BT-2020M-E
Code : 04
Additional Details : Weight : 8.00000kg
Description
- Sealed to allow wet gels on surface
- For use in Syngene Systems
Specifications
UV | |
302 nm | |
100/115/220/230 V |
Fixed | |
Benchtop UV Transilluminator |