missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Synth-a-Freeze™ Cryopreservation Medium

Product Code. 10364330
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10364330

Brand: Gibco™ A1254201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Synth-a-Freeze™ Cryopreservation Medium

Synth-a-Freeze™ is a defined, liquid cryopreservation medium containing 10% dimethylsulfoxide (DMSO) without antibiotics, antimycotics, hormones, growthfactors, serum or protein and buffered with HEPES and sodium bicarbonate. Synth-a-Freeze is intended for freezing and storing a variety of mammalian cell types including human keratinocytes, embryonic stem cells, neural stem cells and mesenchymal stem cells. It should not be used for melanocytes.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Keratinocyte Cell, Embryonic Stem Cell, Neural Stem Cell, Mesenchymal Stem Cell
Classification Animal Origin-free
Form Liquid
Product Type Cryopreservation Media
Serum Level Serum-free
Sterility Sterile
With Additives HEPES, Phenol Red, DMSO (10%)
Product Line Gibco, Synth-a-Freeze
Quantity 50 mL
Shipping Condition Dry Ice
Endotoxin Level < 1 EU/ml
Purity or Quality Grade Qualified for Further Manufacturing
Content And Storage Store in freezer (-5 to -20°C).
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.