missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Synthegel™ 3D hiPSC Grow Mix, 1 mg, Lyophilized Powder
The hiPSC Grow Mix supplement provides a complete environment for hiPSC 3D culture.
Brand: Corning™ 354792
Additional Details : Weight : 0.04300kg
Description
- Matrix rigidity is tunable by altering the peptide concentration
- Purified synthetic peptide in neutral pH
- Cells are encapsulated in physiological mimicking conditions
- Fast hydrogel formation (5 to 30 minutes)
- Sol-gel transfer reversible by shearing force
- Works with standard cell culture methodologies, growth factors and media
- Allows growth and passaging of hiPSCs in 3D format
- The hiPSC Grow Mix supplement provides a complete environment for hiPSC 3D culture
- Allows 3D embedded and suspension hiPSC cultures
Specifications
Store at 2 to 8°C | |
3D Cell Culture | |
Extracellular Matrix | |
hiPSC 3D Culture Matrix Kits | |
Chemically Defined |
1 mg hiPSC Growth Mix Supplement | |
Lyophilized | |
Non-sterile | |
1 mg |