missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Synthegel™ 3D hiPSC Suspension Matrix Kit, 16 mL

Powerful bio-tool for large-scale manufacturing of physiological 3D hiPSC spheroids in a lab setting. After harvesting, spheroids can be used directly for various downstream tasks such as drug screening, bioprinting for tissue engineering, and somatic cell differentiation.

Brand:  Corning™ 354791

Product Code. 17952291

  • £550.00 / 16mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Kit consists of a vial of Synthegel hiPSC suspension matrix, a nanofiber solution for 3D suspension culture and a vial of Synthegel X-Link solution
  • Requires a vial of lyophilized Synthegel 3D hiPSC Grow Mix
  • Matrix rigidity is tunable by altering the peptide concentration
  • Purified synthetic peptide in neutral pH
  • Cells are encapsulated in physiological mimicking conditions
  • Fast hydrogel formation (5 to 30 minutes)
  • Sol-gel transfer reversible by shearing force
  • Works with standard cell culture methodologies, growth factors and media
  • Allows growth and passaging of hiPSCs in 3D format. The hiPSC Grow Mix supplement provides a complete environment for hiPSC 3D culture
  • Allows 3D culture of hiPSC in suspension
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ISO
Store at 2 to 8°C
Synthegel™ 3D hiPSC Suspension Matrix Kit
Liquid
Sterile-filtered
3D suspension culture of hiPSC in a synthetic peptide hydrogel
Hydrogel
Ambient Temperature
Red
Ambient Temperature
3D Cell Culture
Extracellular Matrix
Kit
Neutral
16 mL
Synthetic Peptide Matrix
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers