Learn More
Corning™ Synthegel™ Spheroid Matrix Kit, 10 mL
Research tool for high content 3D biologically formed spheroid culture with more accurate in vivo predictions. The matrix has a stronger gel and optimal cytocompatibility for spheroid culture. The kit consists of a Spheroid matrix peptide nanofiber solution and a Synthegel X-Link solution.
Brand: Corning™ 354789
Additional Details : CAS Number : 7732-18-5 Weight : 0.21500kg
Description
- Matrix rigidity is tunable by altering the peptide concentration
- Purified synthetic peptide in neutral pH
- Cells are encapsulated in physiological mimicking conditions
- Fast hydrogel formation (5 to 30 minutes)
- Sol-gel transfer reversible by shearing force
- Works with standard cell culture methodologies, growth factors, and media
Specifications
ISO | |
Store at 2 to 8°C | |
Synthegel™ Spheroid Matrix Kit | |
Liquid | |
Sterile-filtered | |
Culture of cancer spheroids in a synthetic peptide hydrogel | |
Hydrogel | |
Ambient Temperature |
Clear | |
Ambient Temperature | |
3D Cell Culture | |
Extracellular Matrix | |
Kit | |
Neutral | |
10 mL | |
Synthetic Peptide Matrix |