missing translation for 'onlineSavingsMsg'
Learn More

Novex™ Synthetic Block (10X)

Synthetic Block (10X)

Brand:  Novex™ PA017

Code : 50

Additional Details : Weight : 0.06000kg

Product Code. 10524335

  • £74.50 / Each
Estimated Shipment: 05-08-2024
to see stock.

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Invitrogen’s 10x Synthetic Block is a concentrated polymer-based protein-free blocking agent. This easy to use blocking agent has been optimized for use with Invitrogen’s ProtoArray Human Protein Microarrays and results in optimal assay performance and signal-to-background ratio. Sufficient reagent for processing 10 arrays.
Specifications

Specifications

Synthetic Block (10X), 75mL, 1 Bottle, Blocking/Background Suppression Reagent, Room Temperature, Sufficient Reagent For Processing 10 Arrays, For Protein Assays and Analysis
1 Bottle
75 mL 10X Blocking Agent
Room Temperature
Synthetic Block
Protein Assay and Analysis
10X
6.9 Undiluted, 7.44 diluted 1:10
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.