missing translation for 'onlineSavingsMsg'
Learn More

PURUS™ Synthetic Cleanroom Paper

Product Code. 15723579 Shop All Purus Products
Click to view available options
Color:
Blue
Green
Pink
White
Yellow
Size:
A2
A3
A4
A5
A6
Weight (Metric):
100 gsm
105 gsm
80 gsm
Quantity:
250 Sheets/Pk., 10 Packs/Cs.
250 Sheets/Pk., 20 Packs/Cs.
250 Sheets/Pk., 4 Packs/Cs.
250 Sheets/Pk., 40 Packs/Cs.
250 Sheets/Pk., 5 Packs/Cs.
Unit Size:
Pack of 10
Pack of 1250
Pack of 250
Pack of 5
Pack of 5000
This item is not returnable. View return policy

Product Code. 15723579

Brand: PURUS™ LCIB 2040C

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Cleanroom paper that excels in Tensile Strength. For writing, printing and photocopying in cleanroom environments, PURUS™ Cleanroom Paper provides excellent printability and heat toner adhesion and is recyclable.

  • Features high tensile strength and excellent heat resistance for enhanced copier and printing efficiency
  • Proprietary binding technology results in very low levels of particles, nonvolatile residue (NVR) and ion extractables
  • Unique precision cutting technology guarantees consistent sizing and cleanliness
  • Produced in ISO 4 (Class 10) cleanroom
  • Recyclable
  • Suitable for use in ISO 4 to 5 (Class 10 to Class 1000) cleanrooms

Specifications

Material Synthetic
Ruled No
Color Blue
Dimensions (L x W) 420 x 297 mm
Autoclavable Autoclavable
For Use With (Application) For Class 10 to Class 1000 cleanrooms, writing, printing and photocopying in clean environments
Plain paper Yes
Product Type Cleanroom Paper
Quantity 250 Sheets/Pk., 5 Packs/Cs.
Size A3
Weight (Metric) 80 gsm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.