Learn More
Synthetic Sea Water for ASTM 665 / IP 135, Paragon Scientific™
A high quality analytical reagent manufactured and certified compliant for use in ASTM D 665 / IP 135 the Standard Test Method for Rust-Preventing Characteristics of Inhibited Mineral Oil in the Presence of Water.
£171.00 - £445.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16070420
|
Paragon Scientific
SSWS-30 |
10 x 30 mL |
£171.00
Pack of 10 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16080420
|
Paragon Scientific
SSWS-300 |
300 mL |
£181.00
300mL |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16090420
|
Paragon Scientific
SSWS-500 |
500 mL |
£289.00
500mL |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16000430
|
Paragon Scientific
SSWS-5L |
5 L |
£445.00
5 L |
Estimated Shipment: 14-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- A Synthetic Sea Water Analytical Reagent manufactured in accordance with ASTM D 665 / IP 135.
- Provided with a certificate which includes an estimation of uncertainty of manufacture.
- Ensures full compliance with ASTM and IP test method requirements for reagent preparation.
- Conveniently packaged in single dose or multi dose packs.
- Cost effective and time saving
- Certificate of Manufacture & MSDS included
- 12 month shelf life