missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ SYPRO™ Protein Gel Stains
Liquid format protein gel stains
Brand: Lonza™ 50562
Additional Details : Weight : 1.63290kg
Description
SYPRO Ruby Protein Gel Stains
- Ideal for 2-D gel analysis Sensitivity rivals silver stain Detects nanogram levels of protein Offers a broad linear quantitation range and consistent gel-to-gel staining for accurate protein expression comparisons
- Provides high sensitivity for detection of proteins in SDS-polyacrylamide gels
- Does not require the use of acids or organic solvents
- Suitable for use with saline solutions Ideal for use when eluting active protein complexes from gels because the mild staining conditions are unlikely to affect protein activity
- Detects as little as 4 to 8ng of protein per band or proteins as small as 6.5kDa Provides linear protein quantitation up to three orders of magnitude, from ng to μg per band
- Offers complete staining in less than one hour—no fixation or destaining required
- Compatible with many types of detection systems Recommended for instruments that employ frequency-doubled Nd:YAG or green or red He-Ne lasers
Product licensed by Cambrex from Molecular Probes, Inc. For research and quality control use only.
Specifications
Ruby | |
Liquid | |
4°C |
Protein Stain | |
1 L |