Learn More
Hamilton™ Custom Cemented Needle Syringes
The gas-tight version of the original 700 series syringes.
Brand: Hamilton™ DS80008
This item is not returnable.
View return policy
Description
- Ideal for dispensing volumes from 1 up to 500 μL
- A variety of volumes, lengths, and gauges available
Specifications
Not Autoclavable | |
0.46 mm | |
Gastight | |
50°C | |
10°C | |
10 μL | |
Cemented |
Custom | |
6.60 mm | |
68.9 bar | |
1701 | |
1 Ea. | |
Custom |