Learn More
Hamilton™ 1000 Series Luer Lock Syringes
A mid-volume gastight syringe.
Brand:  Hamilton™ 85021
Product Code. 11523254
  • £389.00 / Each
Explore more special offers
Description
Description
  • 50mL volume
  • For Metal or Kel-F Hubs
  • Without flange
Specifications
Specifications
Not Autoclavable
6.9 bar
1050
50 mL
Syringe Needle
50°C
1/Pk.
Luer Lock