Learn More
Invitrogen™ SYTO™ Green Fluorescent Nucleic Acid Stains
SYTO Green Fluorescent Nucleic Acid Stains exhibit bright, green fluorescence upon binding to nucleic acids. These dyes are cell-permeant and can be used to stain RNA and DNA in live cells, dead cells, and bacteria.
Brand: Invitrogen™ S34854
257.64 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
The SYTO Green Fluorescent Nucleic Acid Stains exhibit bright, green fluorescence upon binding to nucleic acids. These dyes are cell-permeant and can be used to stain RNA and DNA in live cells, dead cells, and bacteria. Because the dye characteristics and staining pattern of the SYTO dyes may vary, we offer 10 different SYTO Green Fluorescent Nucleic Acid Stains available individually along with the SYTO Green Fluorescent Nucleic Acid Stain Sampler Kit #1 (containing SYTO 11–14, 16, 21, and 24) to enable researchers to find the most appropriate green-fluorescent SYTO stain for their system . We also provide the SYTOX Green Nucleic Acid Stain (Cat. No. S7020 and R37109), which is a cell-impermeant green-fluorescent stain used for staining dead cells or performing nuclear counterstaining in fixed cells.
SYTO 9 green fluorescent nucleic acid stain has been shown to stain live and dead Gram-positive and Gram-negative bacteria, and it is a component of the LIVE/DEAD BacLight Bacterial Viability Kits (Cat. No. L-7007, L-7012, L-13152).
Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.
Order Info
Shipping Condition: Room Temperature
Specifications
Green | |
Store in freezer at -5°C to -30°C and protect from light. | |
Cell-Permeant | |
Fluorescence Microscope, High Content Imager, Flow Cytometer | |
100 μL | |
SYTO 9 Green Fluorescent Nucleic Acid Stain | |
SYTO™ | |
Fluorescent Dye | |
Nucleic Acids |
5 mM | |
RNA: 486 nm DNA: 485 nm |
|
Fluorescent Labeling, Live Cell Imaging, Microarray | |
100 μL | |
Fluorescence | |
RNA: 501 nm DNA: 498 nm |
|
Room Temperature | |
Nucleic Acid Stain |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.