missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ SYTOX™ Orange Dead Cell Stain, for flow cytometry

Product Code. 11320972
Click to view available options
Quantity:
1 mL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11320972

Brand: Molecular Probes™ S34861

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

SYTOX™ Orange Dead Cell Stain is a bright, easy-to-use nucleic acid stain for distinguishing dead from live cells in flow cytometry assays.

  • A bright, easy-to-use nucleic acid stain for distinguishing dead from live cells in flow cytometry assays
  • Designed for flow cytometry using the 488 nm blue laser, or 532 nm green laser with emission in the 575/26 BP filter (similar to PE)
  • The dye concentration has been optimized for samples with concentrations ranging from 1 x 104 to 1 x 106 cells/mL
  • Stain has been validated in several cell types including Jurkat, ReH, CHO, Kg1a, 3T3, HeLa, HEK (primary cells), and lysed whole blood
  • It has also been tested in several applications including apoptosis, cell cycle, cell proliferation, and immunophenotyping assays
TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Impermeant
Color Orange
Content And Storage Contains 1 vial of SYTOX™ orange dead cell stain (250 μM solution in DMSO).

Store in freezer (-5°C to -30°C), and protect from light.
Description SYTOX™ Orange Dead Cell Stain, for flow cytometry
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Dead Cell Stain
Dye Type SYTOX™ Orange
Emission 570
Excitation Wavelength Range 547 nm
Form Solution
Format Tube(s)
Product Line SYTOX
Quantity 1 mL
Shipping Condition Room Temperature
Solubility DMSO (Dimethylsulfoxide)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.