Learn More
Thermo Scientific™ T-865 Fixed Angle Rotor for WX+ Ultraspeed Centrifuges
Perform a variety of applications with the Thermo Scientific™ T-865 Fixed Angle Rotor, which offers eight-place capacity.
Brand:  Thermo Scientific™ 51411
21783.11 GBP valid until 2025-06-30
Use promo code "24069" to get your promotional price.
Product Code. 12101690
  • £21783.11 / Each
Explore more special offers
Description
Description
Perform a variety of applications with the Thermo Scientific™ T-865 Fixed Angle Rotor, which offers eight-place capacity.
Recommended for:
Differential protein fractionation, DNA isolation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
T-865 Fixed Angle Rotor
Fixed Angle Rotor
429,459 x g
91 mm
51.7
25 mm
Thermo Scientific™ Sorvall™ WX ultraCentrifuges
305,340 x g
Titanium
8 x 36 mL
65,000 rpm
38.4 mm
97 mm
18.3 lb. (8.3 kg)
23.5°
181,222 x g
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ T-865 Fixed Angle Rotor for WX+ Ultraspeed Centrifuges > T-865 Fixed-angle rotor