missing translation for 'onlineSavingsMsg'
Learn More

t-Butyl4-[4-(4,4,5,5-teramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazinecarboxylate, 90%, Thermo Scientific™

Product Code. 11376825
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 470478-90-1
Molecular Formula C21H33BN2O4
Molecular Weight (g/mol) 388.32
MDL Number MFCD06411544
InChI Key ZMAVVXGWEHZLDW-UHFFFAOYSA-N
Synonym tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine-1-carboxylate, 4-4-boc-piperazino phenylboronic acid pinacol ester, 4-4-n-boc piperazin-1-yl phenylboronic acid pinacol ester, 4-4-tert-butoxycarbonylpiperazinyl phenylboronic acid pinacol ester, 4-4-tert-butoxycarbonylpiperazinyl phenylboronic acid, pinacol ester, tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl tetrahydro-1 2h-pyrazinecarboxylate, 4-4-boc-piperazinyl phenylboronic acid pinacol ester, 4-n-tert-butoxycarbonylpiperazine benzeneboronic acid pinacol ester, tert-butyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine-1-carboxylate
PubChem CID 2795514
IUPAC Name tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 388.31
Chemical Name or Material tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl) phenyl]piperazine-1-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'