Learn More
Grant Instruments™ T-Clamp
The Grant T-Clamp allows a Grant heating circulator to be attached to virtually any vertical sided vessel.
Brand:  Grant Instruments™ T-CLAMP
Product Code. 12717769
  • £102.00 / Each
Estimated Shipment: 21-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
The Grant T-Clamp allows a Grant heating circulator to be attached to virtually any vertical sided vessel.
Grant heated circulators are suitable for use with Grant stainless steel and plastic tanks. With the addition of a clamp (T-Clamp) they can also be attached to virtually any vertical sided tank with a maximum wall thickness of 35mm for rectangular tanks, 30mm for circular tanks (300mm diameter), and a capacity of up to 50 litres. Minimum and maximum temperatures achievable are dependent upon the tank insulation and minimum operating temperature depends on the accessory cooling device.
Specifications
Specifications
50 L
Allows heating circulator to be attached to virtually any vertical sided vessel with a maximum wall