missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ T-PER™ Tissue Protein Extraction Reagent

Product Code. 10730995 Shop All Thermo Scientific Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 10730995

Brand: Thermo Scientific™ 78510

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extract total protein from tissue samples using this mild cell lysis and extraction reagent developed specifically for tissues.

Perform mild extraction of total protein from tissue samples with Thermo Scientific™ T-PER Tissue Protein Extraction Reagent.

This tissue cell lysis reagent utilizes a proprietary detergent in 25mM bicine and 150mM sodium chloride (pH 7.6) to maximize the efficiency of protein solubilization from mammalian tissue samples by homogenization. The simple composition is compatible with additives such as protease inhibitors, salts, reducing agents and chelating agents, providing versatility for many different sample types and lysis applications. Cell lysates prepared with T-PER Reagent are directly compatible with reporter assays (e.g., luciferase, beta-galactosidase, chloramphenicol acetyl transferase), protein kinase assays (e.g., PKA, PKC, tyrosine kinase), immunoassays (e.g., Western blots, ELISAs, RIAs) and/or protein purification procedures.

Highlights:

  • Simple procedure – just homogenize tissue sample in 1:20 (w/v) of tissue to T-PER Reagent, then centrifuge to pellet cell/tissue debris

  • Mild and convenient – detergent is dialyzable for quick and easy removal; can be used with additional components (e.g., protease inhibitors, salts, reducing agents, chelating agents, etc.)

  • Flexible – lysate may be used for reporter assays, protein kinase assays, immunoassays and/or protein purifications

  • Compatible – the lysate is compatible with standard protein assays such as Thermo Scientific Pierce™ Coomassie Plus™ (Bradford) Protein Assay

TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 mL
Format Liquid
Product Type Tissue Protein Extraction Reagent
Volume (Metric) 500 mL
Content And Storage Store at room temperature.
Product Line T-PER

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.