Learn More
Thermo Scientific™ T Phosphoramidite, standard grade, 28-400 finish bottle
T CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxythymidine (dT) for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides.
Brand: Thermo Scientific™ 27-1736-13
13.12 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Description
T CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxythymidine (dT) for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups (except T and U) designed specifically to protect the DNA during the chemical synthesis process, and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. The 'Standard Grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 30 mL Amber bottle with a 28-400 Finish neck style, which is compatible with most MerMade Systems of oligonucleotide synthesizers.
Features:
• Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-2'-deoxythymidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula: C40H49N4O8P
• Base: T
• Formula weight: 744.82 g/mol
• CAS number: 98796-51-1
• HPLC Purity: ≥98.0%
• 31P NMR Purity: ≥98.0%
Specifications
MerMade Synthesizers | |
Store in freezer (at or below -20°C). | |
Room Temperature | |
2 g | |
Standard | |
Powder |
T CEP | |
Deoxythymidine (dT) | |
98% | |
Phosphoramidite | |
DNA |
For Research Use Only. Not for use in diagnostic procedures.