Learn More
GVS T200 Respirator and Head Harness
The RPB T200 enables you to work in your environment free of respiratory hazards and discomfort through its supportive design. Very lightweight and comfortable to help protect the user better. It provides a wide viewing area to make your work easier.
Brand:  GVS 17-200-22-CE
Product Code. 18429645
  • £275.00 / Each
Explore more special offers
Description
Description
The RPB T200 enables you to work in your environment free of respiratory hazards and discomfort through its supportive design.
  • Very lightweight and comfortable to help protect the user better. It provides a wide viewing area to make your work easier.
  • Multi adjustment to fit a wide range of users.
  • Certified to EN 12941 1998 + A2:2008 (TH3) and EN 166 2001 1 F 3
  • Includes: Tychem 2000 face seal, head harness assembly, PX5™ breathing tube
  • To use with PAPR PX5™
Specifications
Specifications
White
PAPR System with Hood and Head Harness
One Size