Learn More
Simport™ Scientific T310 Series Cryovials
T310 Series Cryovials are designed for storing biological material, human or animal cells, these cryovials are compatible with most storage systems.
Specifications
Material Polypropylene
Sterility Sterile
For Use With (Application) For storing biological material, human or animal cells, these cryovials are compatible with most storage systems
Thread Style External
RNase Free Yes
View More Specs
Description
Description
  • Cap features a long skirt for easy one-hand aseptic methods and an exclusive silicone seal fitted inside the cap
  • Super-fast thread design allows removal or sealing with a mere 1 1/4 turn
  • Inside thread design that will not contribute to possible contamination
  • Tubes are provided with a white marking area for sample identification
  • Sterilized by gamma radiation
  • Autoclavable
  • Polypropylene construction
Specifications
Specifications
Polypropylene
For storing biological material, human or animal cells, these cryovials are compatible with most storage systems
Yes
Autoclavable
Yes
Sterile
External
Yes
Cryogenic Vial