missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ T4 DNA Ligase (5 U/μL)

T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5'-phosphate and 3'-hydroxyl termini in duplex DNA or RNA.

Brand:  Thermo Scientific™ EL0011

45.79 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 10723941

  • £65.50 / 1000 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

5 U/μL
10X T4 DNA Ligase Buffer
T4 DNA Ligase
DNA Ligase
1,000 units
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2817470 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberEL0011
Lot Number2806278 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberEL0011
Lot Number2833808 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberEL0011
Lot Number2833831 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberEL0011
Lot Number2833816 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberEL0011
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ T4 DNA Ligase (5 U/μL) > T4 DNA ligase; 5U/uL; 1000U

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.