missing translation for 'onlineSavingsMsg'
Learn More
Learn More
T7 Epitope Tag Antibody, Agarose, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Bio-Techne NB600-378
Additional Details : Weight : 0.00970kg
Description
T7 Epitope Tag Polyclonal antibody specifically detects T7 Epitope Tag in Epitope Tag samples. It is validated for Immunoprecipitation.Specifications
T7 Epitope Tag | |
Polyclonal | |
Agarose | |
PBS with 0.09% Sodium Azide | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
IgG |
Immunoprecipitation | |
0.25 mg/mL | |
Immunoprecipitation 1:10-1:500 | |
MASMTGGQQMG epitope tag, MASMTGGQQMG tag, T7 epitope tag | |
Goats were immunized with T7 cleavage site (MASMTGGQQMG) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support. | |
0.1 mg | |
Epitope Tags, T7 Epitope Tags | |
Store at 4C. Do not freeze. | |
Epitope Tag |
For Research Use Only