missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TA Cloning™ Kit, Dual Promoter, with pCR™II Vector and One Shot™ INVαF' Chemically Competent E. coli
Provides quick, one-step cloning strategy for directly inserting a Taq-amplified PCR product into plasmid vector
£388.00 - £650.00
Specifications
Bacterial or Yeast Strain | INVαF ́ |
---|---|
Cell Type | Chemically Competent E. coli |
Cloning Method | TA Cloning |
Content And Storage | This kit contains linearized pCR™II vector, ExpressLink™ T4 DNA ligase, 5X ExpressLink™ T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, and controls. Competent cell kits contain One Shot™ chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid. Store One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored. |
Format | Kit |
Product Code | Brand | Bacterial or Yeast Strain | Includes | No. of Reactions | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bacterial or Yeast Strain | Includes | No. of Reactions | Price | Quantity & Availability | |||
10003102
|
Invitrogen™
K205001 |
INVαF ́ | Linearized pCRII vector, ExpressLink T4 DNA ligase, 5X ExpressLink T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, controls, One Shot INVαF' chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid. | 20 Reactions |
£388.00
Each |
Estimated Shipment: 15-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10740165
|
Invitrogen™
K205040 |
INVαF ́ | Linearized pCRII vector, ExpressLink T4 DNA ligase, 5X ExpressLink T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, controls, One Shot INVαF' chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid. | 40 Reactions |
£650.00
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
The T7 and Sp6 promoters of the pCRII vector allow in vitro transcription of the insert to produce sense or anti-sense products. The TA Cloning Kit Dual Promoter uses the pCRII cloning vector and ExpressLink T4 DNA Ligase to generate a ligation product in a fifteen-minute, room-temperature ligation step. Reactions typically yield >80% recombinants containing inserts.
- Fast and convenient: 15-minute, room-temperature ligation
- Efficient: blue/white screening and >80% clones with correct insert
- Flexible: choice of kanamycin or ampicillin resistance for flexible antibiotic selection
- Hassle-free: eliminates any enzymatic modifications of PCR product
- Streamlined: does not require use of PCR primers that contain restriction sites
- 3'-T overhangs for direct ligation of Taq-amplified PCR products
- T7 and Sp6 promoters for in vitro RNA transcription and sequencing
- Versatile polylinker with flanking EcoR I sites for easy excision of inserts
- M13 forward and reverse primer sites for sequencing
- How TA Cloning Works\
- Taq polymerase has non-template-dependent activity that adds single deoxyadenosine (A) to 3' ends of PCR products
- Linearized vector supplied in this kit has single 3' deoxythymidine (T) residues
- Allows PCR inserts to ligate efficiently with vector
- Kit Configurations
- TA Cloning Kit is offered in a variety of configurations: with One Shot INVF' Chemically Competent E. coli (K2050-01 and K2050-40), One Shot TOP10F' Chemically Competent E. coli (K2060-01 and K2060-40), and without competent cells (K2750-20 and K2750-40) in 20- and 40-reaction kit sizes
Cloning, PCR Cloning
Specifications
INVαF ́ | |
TA Cloning | |
Kit | |
Linearized pCRII vector, ExpressLink T4 DNA ligase, 5X ExpressLink T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, controls, One Shot INVαF' chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid. | |
Cloning Kit | |
pCRII |
Chemically Competent E. coli | |
This kit contains linearized pCR™II vector, ExpressLink™ T4 DNA ligase, 5X ExpressLink™ T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, and controls. Competent cell kits contain One Shot™ chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid. Store One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored. |
|
PCR Cloning | |
One Shot™ | |
T7, SP6 |
For Research Use Only. Not for use in diagnostic procedures.