Learn More
Sartorius Tacta™ Mechanical Pipettes
GREENER_CHOICEBIOPHARMA
Product Code. 15516314 Shop All Sartorius Products
Click to view available options
Number of Channels:
1
8
12
Volume (Metric):
0.1 to 3 μL
0.5 to 10 μL
10 to 100 μL
100 to 1000 μL
1000 to 10,000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
5 to 100 μL
500 to 5000 μL
Color:
Blue
Gray
Green
Orange
Red
Yellow
Unit Size:
Each
Product Code. 15516314
Brand: Sartorius LH729070
Only null left
Add to basket
Add to basket
Exceptionally balanced design is effortless to use and exceptionally comfortable to hold
  • Comfortable to hold due to the ergonomically designed handle
  • Unique Sartorius Optilock system provides flexibility for volume adjustment and locking
  • Controlled and smooth tip ejection with the new Sartorius Optiject technology
  • Calibration adjustment to provide accurate results for various liquid types
  • Easy to clean, with only three parts to disassemble
  • Reliable results, even over lengthy pipetting periods
TRUSTED_SUSTAINABILITY
Specifications
Compatible Tips Optifit 1000 μL, SafetySpace Filter Tips 1000 μL
Adjustable Tip Spacing Yes
Autoclavable Autoclavable
Color Blue
Disposable Reusable
Grip Type Ergonomic
Material Polypropylene
Number of Channels 1
Product Line Tacta
Programmability No
Sterility Non-sterile
Type Pipette
Volume (Metric) 100 to 1000 μL
Color Code Yellow