Learn More
Sartorius Tacta™ Mechanical Pipettes
GREENER_CHOICEBIOPHARMA
Exceptionally balanced design is effortless to use and exceptionally comfortable to hold
Brand:  Sartorius LH-729080
336.79 GBP valid until 2024-12-31
Use promo code "21925" to get your promotional price.
 View more versions of this product
Product Code. 15526314
  • £487.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Comfortable to hold due to the ergonomically designed handle
  • Unique Sartorius Optilock system provides flexibility for volume adjustment and locking
  • Controlled and smooth tip ejection with the new Sartorius Optiject technology
  • Calibration adjustment to provide accurate results for various liquid types
  • Easy to clean, with only three parts to disassemble
  • Reliable results, even over lengthy pipetting periods
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Optifit Tips: 5000μL
Green
Tacta™
500 to 5000 μL
Autoclavable
1
Pipette
Green
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products