missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TADA1L Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20337-1-AP-20UL
This item is not returnable.
View return policy
Description
The MW of this protein is 37 kDa, and 20337-1-AP specifically recognizes the 37 kDa protein.
Probably involved in transcriptional regulation.Specifications
TADA1L | |
Polyclonal | |
Unconjugated | |
TADA1 | |
KIAA0764, RP1 9E21.4, SPT3 associated factor 42, STAF42, TADA1, TADA1L, Transcriptional adapter 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117143, 27878 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence | |
0.59 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96BN2, Q99LM9 | |
Tada1 | |
TADA1L Fusion Protein Ag14118 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction